Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASP5
Entrez Gene ID
838
Gene Name
caspase 5
Gene Aliases
ICE(rel)III, ICEREL-III, ICH-3
Chromosome Location
Chr.11: 104994240 - 105023902 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136109.1 NP_001129581.1 2 - 3 295 143
NM_001136112.1 NP_001129584.1 3 - 4 508 143
NM_004347.3 NP_004338.3 3 - 4 469 143
XM_011543021.2 XP_011541323.1 3 - 4 1203 143
GenBank mRNA AK296660.1 - 2 - 3 294 143
BC074994.2 - 2 - 3 414 143
DQ228672.1 - 2 - 3 437 143
DQ228673.1 - 1 - 2 263 143
DQ228677.1 - 2 - 3 476 143
DR005081.1 - 2 - 3 523 143
U28015.1 - 2 - 3 423 143
X94993.1 - 2 - 3 330 143

View Full Product Details