The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006196.3 | NP_006187.2 | 1 - 1 | 566 | 115 |
GenBank mRNA | AK130439.1 | - | 1 - 1 | 247 | 115 |
AK314225.1 | - | 1 - 1 | 433 | 115 | |
BC039742.1 | - | 1 - 1 | 427 | 115 | |
FJ224329.1 | - | 1 - 1 | 427 | 115 | |
U24223.1 | - | 1 - 1 | 452 | 115 | |
X78137.1 | - | 1 - 1 | 305 | 115 | |
Z29505.1 | - | 1 - 1 | 360 | 115 |