Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50580251-50580745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHKB
Entrez Gene ID
1120
Gene Name
choline kinase beta
Gene Aliases
CHETK, CHKL, CK, CKB, CKEKB, EK, EKB, MDCMC
UniGene
Hs.654827
Gene Chromosome Location
Chr.22: 50578958-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Gene Symbol
CHKB-CPT1B
Entrez Gene ID
386593
Gene Name
CHKB-CPT1B readthrough (NMD candidate)
Gene Aliases
CHKL-CPT1B
UniGene
Hs.439777
Gene Chromosome Location
Chr.22: 50568861-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5819246 c.678-2A>G p.? Chr.22: 50580418-50580418 on Build GRCh38
4993903 c.670A>C p.N224H Chr.22: 50580572-50580572 on Build GRCh38
224619 c.582-1G>A p.? Chr.22: 50580661-50580661 on Build GRCh38
4445643 c.582-3C>T p.? Chr.22: 50580663-50580663 on Build GRCh38
5989449 c.704C>T p.P235L Chr.22: 50580390-50580390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764281449 Chr.22: 50580533-50580533 on Build GRCh38
rs6010012 Chr.22: 50580513-50580513 on Build GRCh38
rs765702456 Chr.22: 50580281-50580281 on Build GRCh38
rs767551132 Chr.22: 50580309-50580309 on Build GRCh38
rs868600295 Chr.22: 50580637-50580637 on Build GRCh38
rs747744641 Chr.22: 50580418-50580418 on Build GRCh38
rs780859142 Chr.22: 50580700-50580700 on Build GRCh38
rs192728067 Chr.22: 50580278-50580278 on Build GRCh38
rs766375396 Chr.22: 50580525-50580525 on Build GRCh38
rs552898356 Chr.22: 50580623-50580623 on Build GRCh38
rs997355420 Chr.22: 50580518-50580518 on Build GRCh38
rs370210722 Chr.22: 50580465-50580465 on Build GRCh38
rs768970529 Chr.22: 50580604-50580604 on Build GRCh38
rs746452829 Chr.22: 50580693-50580693 on Build GRCh38
rs758344083 Chr.22: 50580399-50580399 on Build GRCh38
rs761978769 Chr.22: 50580612-50580612 on Build GRCh38
rs375851386 Chr.22: 50580452-50580452 on Build GRCh38
rs766718421 Chr.22: 50580384-50580384 on Build GRCh38
rs751522981 Chr.22: 50580636-50580636 on Build GRCh38
rs767355484 Chr.22: 50580659-50580659 on Build GRCh38
rs769300409 Chr.22: 50580419-50580419 on Build GRCh38
rs768336190 Chr.22: 50580344-50580344 on Build GRCh38
rs1024231997 Chr.22: 50580703-50580703 on Build GRCh38
rs199783402 Chr.22: 50580402-50580402 on Build GRCh38
rs1049720704 Chr.22: 50580603-50580603 on Build GRCh38
rs752410851 Chr.22: 50580543-50580543 on Build GRCh38
rs779762736 Chr.22: 50580400-50580400 on Build GRCh38
rs753767029 Chr.22: 50580677-50580678 on Build GRCh38
rs747689391 Chr.22: 50580365-50580365 on Build GRCh38
rs888546734 Chr.22: 50580629-50580629 on Build GRCh38
rs773724394 Chr.22: 50580383-50580383 on Build GRCh38
rs763052761 Chr.22: 50580459-50580461 on Build GRCh38
rs752801856 Chr.22: 50580313-50580313 on Build GRCh38
rs765505160 Chr.22: 50580336-50580337 on Build GRCh38
rs539357838 Chr.22: 50580347-50580347 on Build GRCh38
rs775848217 Chr.22: 50580428-50580429 on Build GRCh38
rs199847760 Chr.22: 50580390-50580390 on Build GRCh38
rs777309411 Chr.22: 50580549-50580549 on Build GRCh38
rs776677585 Chr.22: 50580607-50580607 on Build GRCh38
rs150676350 Chr.22: 50580389-50580389 on Build GRCh38
rs763332214 Chr.22: 50580288-50580288 on Build GRCh38
rs75187587 Chr.22: 50580684-50580684 on Build GRCh38
rs751708599 Chr.22: 50580554-50580554 on Build GRCh38
rs770306965 Chr.22: 50580570-50580570 on Build GRCh38
rs779897037 Chr.22: 50580338-50580338 on Build GRCh38
rs565326245 Chr.22: 50580593-50580593 on Build GRCh38
rs751732782 Chr.22: 50580302-50580302 on Build GRCh38
rs1051566120 Chr.22: 50580719-50580719 on Build GRCh38
rs749813559 Chr.22: 50580428-50580428 on Build GRCh38
rs370130761 Chr.22: 50580696-50580696 on Build GRCh38
rs993005402 Chr.22: 50580662-50580662 on Build GRCh38
rs780606441 Chr.22: 50580706-50580706 on Build GRCh38
rs776195737 Chr.22: 50580274-50580274 on Build GRCh38
rs372742172 Chr.22: 50580621-50580621 on Build GRCh38
rs184254424 Chr.22: 50580385-50580385 on Build GRCh38
rs772567324 Chr.22: 50580577-50580577 on Build GRCh38
rs200085590 Chr.22: 50580676-50580676 on Build GRCh38
rs745748373 Chr.22: 50580329-50580329 on Build GRCh38
rs368877893 Chr.22: 50580672-50580672 on Build GRCh38
rs114810820 Chr.22: 50580709-50580709 on Build GRCh38
rs1034589454 Chr.22: 50580644-50580644 on Build GRCh38
rs917668820 Chr.22: 50580666-50580666 on Build GRCh38
rs754693809 Chr.22: 50580407-50580407 on Build GRCh38
rs761029141 Chr.22: 50580600-50580600 on Build GRCh38
rs778217852 Chr.22: 50580688-50580688 on Build GRCh38
rs777082533 Chr.22: 50580285-50580286 on Build GRCh38
rs780901838 Chr.22: 50580537-50580537 on Build GRCh38
rs149858290 Chr.22: 50580572-50580572 on Build GRCh38
rs764209313 Chr.22: 50580393-50580393 on Build GRCh38
rs145253375 Chr.22: 50580526-50580526 on Build GRCh38
rs74846249 Chr.22: 50580713-50580713 on Build GRCh38
rs771443755 Chr.22: 50580435-50580435 on Build GRCh38
rs764329647 Chr.22: 50580517-50580517 on Build GRCh38
rs371751084 Chr.22: 50580372-50580372 on Build GRCh38
rs369606974 Chr.22: 50580574-50580574 on Build GRCh38
rs141934594 Chr.22: 50580386-50580386 on Build GRCh38
rs757369551 Chr.22: 50580644-50580644 on Build GRCh38
rs786205118 Chr.22: 50580564-50580564 on Build GRCh38
rs774613929 Chr.22: 50580450-50580450 on Build GRCh38
rs376879066 Chr.22: 50580303-50580303 on Build GRCh38
rs773669615 Chr.22: 50580287-50580287 on Build GRCh38
rs367644242 Chr.22: 50580325-50580325 on Build GRCh38
rs777430309 Chr.22: 50580678-50580678 on Build GRCh38
rs144789175 Chr.22: 50580305-50580305 on Build GRCh38
rs2269382 Chr.22: 50580482-50580482 on Build GRCh38
rs963611942 Chr.22: 50580609-50580609 on Build GRCh38
rs775986184 Chr.22: 50580589-50580589 on Build GRCh38
rs776158594 Chr.22: 50580356-50580356 on Build GRCh38
rs997259215 Chr.22: 50580601-50580601 on Build GRCh38
rs754819467 Chr.22: 50580528-50580528 on Build GRCh38
rs772515839 Chr.22: 50580464-50580464 on Build GRCh38
rs766710559 Chr.22: 50580289-50580289 on Build GRCh38
rs780848949 Chr.22: 50580411-50580411 on Build GRCh38
rs373451619 Chr.22: 50580522-50580522 on Build GRCh38
rs373292797 Chr.22: 50580673-50580673 on Build GRCh38
rs1032658314 Chr.22: 50580720-50580720 on Build GRCh38
rs543829328 Chr.22: 50580569-50580569 on Build GRCh38
rs759899367 Chr.22: 50580293-50580295 on Build GRCh38
rs766816072 Chr.22: 50580671-50580673 on Build GRCh38
rs139952595 Chr.22: 50580392-50580392 on Build GRCh38
rs757296475 Chr.22: 50580318-50580318 on Build GRCh38
rs572515697 Chr.22: 50580477-50580477 on Build GRCh38
rs146409721 Chr.22: 50580645-50580645 on Build GRCh38
rs1015463711 Chr.22: 50580667-50580667 on Build GRCh38
rs755746798 Chr.22: 50580547-50580547 on Build GRCh38
rs961307091 Chr.22: 50580285-50580285 on Build GRCh38
rs750454672 Chr.22: 50580398-50580398 on Build GRCh38
rs772702879 Chr.22: 50580422-50580422 on Build GRCh38
rs577904303 Chr.22: 50580340-50580340 on Build GRCh38
rs1038954782 Chr.22: 50580493-50580494 on Build GRCh38
rs555772646 Chr.22: 50580334-50580334 on Build GRCh38

View Full Product Details