The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014159.6 | NP_054878.5 | 3 - 4 | 4509 | 109 |
XM_011533631.2 | XP_011531933.1 | 4 - 5 | 6005 | 109 | |
XM_011533632.2 | XP_011531934.1 | 3 - 4 | 4581 | 109 | |
XM_011533633.2 | XP_011531935.1 | 4 - 5 | 6007 | 109 | |
XM_011533634.1 | XP_011531936.1 | 3 - 4 | 4534 | 109 | |
XM_017006270.1 | XP_016861759.1 | 4 - 5 | 6008 | 109 | |
GenBank mRNA | AB051519.2 | - | 1 - 2 | 2509 | 109 |
AF161554.1 | - | 1 - 2 | 253 | 109 | |
AJ238403.1 | - | 1 - 2 | 3356 | 109 | |
AY576987.1 | - | 1 - 2 | 3018 | 109 | |
AY576988.1 | - | 1 - 2 | 3018 | 109 | |
BC090954.1 | - | 1 - 2 | 2299 | 109 | |
BC117162.1 | - | 1 - 2 | 3132 | 109 | |
BC117164.1 | - | 1 - 2 | 3132 | 109 |