The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026770.1 | 1 - 1 | 713 | 70 | |
GenBank mRNA | AF143236.1 | - | 1 - 1 | 726 | 70 |
DA188505.1 | - | 1 - 1 | 70 |