The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172766.2 | NP_001166237.1 | 6 - 7 | 1148 | 105 |
NM_001172767.2 | NP_001166238.1 | 6 - 7 | 862 | 105 | |
NM_014491.3 | NP_055306.1 | 6 - 7 | 1151 | 105 | |
NM_148898.3 | NP_683696.2 | 7 - 8 | 1226 | 105 | |
NM_148899.3 | NP_683697.2 | 5 - 6 | 787 | 105 | |
NM_148900.3 | NP_683698.2 | 7 - 8 | 1202 | 105 | |
NR_033766.1 | 10 - 11 | 1364 | 105 | ||
NR_033767.1 | 7 - 8 | 1209 | 105 | ||
XM_017012801.1 | XP_016868290.1 | 9 - 10 | 1832 | 105 | |
GenBank mRNA | AF337817.1 | - | 6 - 7 | 927 | 105 |
AF467253.1 | - | 5 - 6 | 789 | 105 | |
AF467257.1 | - | 6 - 7 | 829 | 105 | |
AF467258.1 | - | 5 - 6 | 774 | 105 | |
AF467259.1 | - | 6 - 7 | 849 | 105 | |
AK131266.1 | - | 10 - 11 | 1348 | 105 | |
AK296957.1 | - | 7 - 8 | 1082 | 105 | |
AY144615.1 | - | 7 - 8 | 1221 | 105 | |
BC126104.1 | - | 7 - 8 | 883 | 105 | |
BC143866.1 | - | 7 - 8 | 883 | 105 | |
BC143867.1 | - | 6 - 7 | 822 | 105 | |
CR749236.1 | - | 12 - 13 | 1377 | 105 | |
DQ778626.1 | - | 3 - 4 | 501 | 105 | |
KJ534839.1 | - | 6 - 7 | 852 | 105 | |
KJ535079.1 | - | 5 - 6 | 990 | 105 | |
KJ535086.1 | - | 5 - 6 | 776 | 105 |