The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018037.1 | NP_001018047.1 | 4 - 5 | 538 | 101 |
NM_001018038.2 | NP_001018048.1 | 4 - 5 | 538 | 101 | |
NM_015186.3 | NP_056001.1 | 4 - 5 | 538 | 101 | |
NM_033305.2 | NP_150648.2 | 4 - 5 | 538 | 101 | |
GenBank mRNA | AB054005.1 | - | 4 - 5 | 449 | 101 |
AF337532.1 | - | 4 - 5 | 538 | 101 | |
AJ608769.1 | - | 4 - 5 | 538 | 101 | |
AJ626859.1 | - | 4 - 5 | 538 | 101 |