The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015356.4 | NP_056171.3 | 28 - 29 | 3921 | 59 |
NM_182706.4 | NP_874365.3 | 28 - 29 | 3921 | 59 | |
GenBank mRNA | AF240677.1 | - | 28 - 29 | 3914 | 59 |
AF271734.2 | - | 28 - 29 | 3892 | 59 | |
AY062238.1 | - | 28 - 29 | 3923 | 59 | |
BC009490.2 | - | 4 - 5 | 301 | 59 | |
BC014632.2 | - | 10 - 11 | 1520 | 59 | |
BC044627.1 | - | 13 - 14 | 1803 | 59 | |
D63481.3 | - | 28 - 29 | 3916 | 59 |