The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002755.2 | NP_001002755.1 | 4 - 5 | 573 | 150 |
NM_015700.3 | NP_056515.2 | 4 - 5 | 663 | 150 | |
XM_017003808.1 | XP_016859297.1 | 4 - 5 | 376 | 150 | |
GenBank mRNA | AF132967.1 | - | 4 - 5 | 375 | 153 |
AJ132584.1 | - | 4 - 5 | 451 | 150 | |
AK300700.1 | - | 4 - 5 | 382 | 150 | |
AK314004.1 | - | 4 - 5 | 406 | 150 | |
AY286306.1 | - | 4 - 5 | 420 | 150 | |
AY286307.1 | - | 4 - 5 | 510 | 150 | |
AY335194.1 | - | 4 - 5 | 566 | 150 | |
BC113692.1 | - | 4 - 5 | 535 | 150 | |
BC113694.1 | - | 4 - 5 | 535 | 150 | |
BU507991.1 | - | 4 - 5 | 378 | 150 | |
BX538347.1 | - | 4 - 5 | 438 | 150 |