The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166108.1 | NP_001159580.1 | 18 - 19 | 3269 | 85 |
NM_001166109.1 | NP_001159581.1 | 16 - 17 | 2383 | 85 | |
NM_001166110.1 | NP_001159582.1 | 9 - 10 | 1882 | 85 | |
NM_016081.3 | NP_057165.3 | 17 - 18 | 3218 | 85 | |
XM_005262861.4 | XP_005262918.1 | 19 - 20 | 3968 | 85 | |
XM_005262866.2 | XP_005262923.1 | 18 - 19 | 3518 | 85 | |
XM_011531768.2 | XP_011530070.1 | 20 - 21 | 4222 | 85 | |
XM_011531769.2 | XP_011530071.1 | 19 - 20 | 4171 | 85 | |
XM_011531770.2 | XP_011530072.1 | 20 - 21 | 4222 | 85 | |
XM_011531771.2 | XP_011530073.1 | 18 - 19 | 3949 | 85 | |
XM_011531772.2 | XP_011530074.1 | 17 - 18 | 3850 | 85 | |
XM_011531773.1 | XP_011530075.1 | 19 - 20 | 3550 | 85 | |
XM_011531774.1 | XP_011530076.1 | 18 - 19 | 3499 | 85 | |
XM_011531775.1 | XP_011530077.1 | 18 - 19 | 2903 | 85 | |
XM_017007910.1 | XP_016863399.1 | 19 - 20 | 4171 | 85 | |
GenBank mRNA | AB023209.1 | - | 9 - 10 | 1846 | 85 |
AF077041.1 | - | 5 - 6 | 1332 | 85 | |
AF151909.1 | - | 5 - 6 | 1238 | 85 | |
AF464873.1 | - | 17 - 18 | 3218 | 85 | |
AK000845.1 | - | 5 - 6 | 807 | 85 | |
AK025843.1 | - | 5 - 6 | 809 | 85 | |
AK095512.1 | - | 16 - 17 | 2345 | 85 | |
BC013867.2 | - | 8 - 9 | 1062 | 85 | |
BC140850.1 | - | 17 - 18 | 3180 | 85 | |
BC144666.1 | - | 18 - 19 | 3231 | 85 | |
BX537391.1 | - | 1 - 2 | 120 | 85 |