The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109878.1 | NP_001103348.1 | 5 - 6 | 771 | 105 |
NM_001109879.1 | NP_001103349.1 | 5 - 6 | 767 | 105 | |
NM_001303475.1 | NP_001290404.1 | 3 - 4 | 525 | 105 | |
NM_016954.2 | NP_058650.1 | 4 - 5 | 664 | 105 | |
XM_005262136.2 | XP_005262193.1 | 5 - 6 | 799 | 105 | |
XM_006724657.3 | XP_006724720.1 | 5 - 6 | 800 | 105 | |
XM_011530972.1 | XP_011529274.1 | 4 - 5 | 1404 | 105 | |
GenBank mRNA | AF251684.1 | - | 3 - 4 | 516 | 105 |
AK097514.1 | - | 5 - 6 | 767 | 105 | |
AY035371.1 | - | 4 - 5 | 643 | 105 | |
BC014194.2 | - | 4 - 5 | 664 | 105 |