The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018064.3 | NP_060534.1 | 4 - 5 | 1181 | 92 |
GenBank mRNA | AK222701.1 | - | 4 - 5 | 1146 | 92 |
BC000764.2 | - | 4 - 5 | 1122 | 92 | |
BC003042.1 | - | 4 - 5 | 1122 | 92 | |
BC005051.1 | - | 4 - 5 | 859 | 92 |