Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TRERF1
- Entrez Gene ID
-
55809
- Gene Name
- transcriptional regulating factor 1
- Gene Aliases
-
BCAR2, HSA277276, RAPA, TREP132, TReP-132, dJ139D8.5
- Chromosome Location
-
Chr.6: 42224933 - 42452216 on Build GRCh38
View Full Product Details