The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020461.3 | NP_065194.2 | 4 - 5 | 1803 | 85 |
GenBank mRNA | AB051456.1 | - | 4 - 5 | 1197 | 85 |
AF272887.1 | - | 4 - 5 | 1396 | 85 | |
AL832617.2 | - | 4 - 5 | 1824 | 85 | |
BC144488.1 | - | 4 - 5 | 1395 | 85 | |
BC144489.1 | - | 4 - 5 | 1395 | 85 | |
BC150613.1 | - | 4 - 5 | 1395 | 85 | |
BC151209.1 | - | 4 - 5 | 1785 | 85 |