The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020461.3 | NP_065194.2 | 23 - 24 | 5680 | 63 |
GenBank mRNA | AF272887.1 | - | 23 - 24 | 5273 | 63 |
AL137665.1 | - | 15 - 16 | 3732 | 63 | |
BC023573.2 | - | 9 - 10 | 2740 | 63 | |
BC144488.1 | - | 23 - 24 | 5248 | 63 | |
BC144489.1 | - | 22 - 23 | 5507 | 63 | |
BC150613.1 | - | 23 - 24 | 5248 | 63 | |
BC151209.1 | - | 23 - 24 | 5721 | 63 |