Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAPER
Entrez Gene ID
49855
Gene Name
S-phase cyclin A associated protein in the ER
Gene Aliases
MSTP063, ZNF291, Zfp291
Chromosome Location
Chr.15: 76348185 - 76905455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145923.1 NP_001139395.1 18 - 19 2445 80
NM_020843.2 NP_065894.2 17 - 18 2302 80
XM_005254417.2 XP_005254474.1 18 - 19 2477 80
XM_005254419.2 XP_005254476.1 18 - 19 2459 80
XM_011521650.1 XP_011519952.1 19 - 20 2562 80
XM_011521652.2 XP_011519954.1 17 - 18 2273 80
XM_011521653.2 XP_011519955.1 17 - 18 2357 80
XM_011521654.2 XP_011519956.1 11 - 12 1621 80
XM_011521656.2 XP_011519958.1 4 - 5 478 80
XM_017022266.1 XP_016877755.1 18 - 19 2482 80
XM_017022267.1 XP_016877756.1 18 - 19 2464 80
XM_017022268.1 XP_016877757.1 18 - 19 2425 80
XM_017022269.1 XP_016877758.1 18 - 19 2457 80
XM_017022270.1 XP_016877759.1 19 - 20 2615 80
XM_017022271.1 XP_016877760.1 18 - 19 2483 80
XM_017022272.1 XP_016877761.1 18 - 19 2477 80
XM_017022273.1 XP_016877762.1 17 - 18 2339 80
XM_017022274.1 XP_016877763.1 18 - 19 2482 80
XM_017022275.1 XP_016877764.1 17 - 18 2314 80
XM_017022276.1 XP_016877765.1 15 - 16 2169 80
XM_017022277.1 XP_016877766.1 17 - 18 2296 80
XM_017022278.1 XP_016877767.1 16 - 17 2178 80
XM_017022279.1 XP_016877768.1 18 - 19 2479 80
XM_017022280.1 XP_016877769.1 18 - 19 2479 80
XM_017022281.1 XP_016877770.1 18 - 19 2479 80
XM_017022282.1 XP_016877771.1 18 - 19 2478 80
XM_017022283.1 XP_016877772.1 18 - 19 2459 80
GenBank mRNA AB040887.1 - 17 - 18 2307 80
AF119814.1 - 11 - 12 1509 80
AF242528.1 - 17 - 18 2277 80
AK025663.1 - 2 - 3 180 80
AK056095.1 - 18 - 19 2466 80
AK056515.1 - 20 - 21 2586 80
BX647285.1 - 18 - 19 2445 80

View Full Product Details