The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020857.2 | NP_065908.1 | 2 - 3 | 571 | 62 |
XM_017022447.1 | XP_016877936.1 | 2 - 3 | 591 | 62 | |
GenBank mRNA | AB040908.1 | - | 2 - 3 | 546 | 62 |
AF308802.1 | - | 2 - 3 | 546 | 62 | |
AL713725.1 | - | 2 - 3 | 571 | 62 | |
BC001513.2 | - | 2 - 3 | 398 | 62 |