The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024529.4 | NP_078805.3 | 5 - 6 | 666 | 85 |
XM_006711537.3 | XP_006711600.1 | 5 - 6 | 590 | 85 | |
GenBank mRNA | AF312865.1 | - | 5 - 6 | 431 | 85 |
AK226038.1 | - | 5 - 6 | 563 | 85 | |
AK300929.1 | - | 5 - 6 | 525 | 85 | |
AK314772.1 | - | 5 - 6 | 424 | 85 | |
BC007325.2 | - | 5 - 6 | 449 | 85 | |
BC014351.1 | - | 1 - 2 | 57 | 85 | |
BC065037.1 | - | 5 - 6 | 503 | 85 | |
BM471846.1 | - | 5 - 6 | 619 | 85 |