Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXL6
Entrez Gene ID
26233
Gene Name
F-box and leucine rich repeat protein 6
Gene Aliases
FBL6, FBL6A, PP14630
Chromosome Location
Chr.8: 144355428 - 144358523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012162.3 NP_036294.2 6 - 7 1068 65
NM_024555.5 NP_078831.4 6 - 7 1050 65
GenBank mRNA AF174592.1 - 6 - 7 725 65
AF370422.1 - 5 - 6 2381 65
AK026541.1 - 6 - 7 999 65
AK223088.1 - 6 - 7 999 65
AL713783.1 - 2 - 3 1279 65
BC033066.1 - 6 - 7 1057 65

View Full Product Details