The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267863.1 | NP_001254792.1 | 1 - 2 | 141 | 75 |
NM_001267864.1 | NP_001254793.1 | 1 - 2 | 141 | 75 | |
NM_031210.5 | NP_112487.1 | 1 - 2 | 141 | 75 | |
GenBank mRNA | AF253980.1 | - | 1 - 2 | 112 | 75 |
AF271779.1 | - | 1 - 2 | 136 | 75 | |
AJ272055.1 | - | 1 - 2 | 100 | 75 | |
AY860853.1 | - | 1 - 2 | 100 | 75 | |
BC017895.1 | - | 1 - 2 | 109 | 75 | |
BP289922.1 | - | 1 - 2 | 113 | 75 | |
BQ083727.1 | - | 1 - 2 | 132 | 75 | |
BU944910.1 | - | 1 - 2 | 141 | 75 | |
BU955571.1 | - | 1 - 2 | 112 | 75 | |
EL951262.1 | - | 1 - 2 | 101 | 75 |