The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160300.1 | NP_001153772.1 | 2 - 3 | 1695 | 97 |
NM_052839.3 | NP_443071.2 | 2 - 3 | 1695 | 97 | |
NR_027691.1 | 3 - 4 | 1746 | 97 | ||
GenBank mRNA | AF398510.1 | - | 2 - 3 | 1665 | 97 |
AF398511.1 | - | 3 - 4 | 1716 | 97 | |
AK096019.1 | - | 2 - 3 | 1679 | 97 | |
AK123508.1 | - | 1 - 2 | 485 | 97 | |
AK299910.1 | - | 2 - 3 | 1695 | 97 | |
BC101023.2 | - | 5 - 6 | 1574 | 97 | |
CR456467.1 | - | 1 - 2 | 1345 | 97 |