Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSGA13
Entrez Gene ID
114960
Gene Name
testis specific 13
Gene Aliases
-
Chromosome Location
Chr.7: 130668646 - 130687432 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304968.1 NP_001291897.1 5 - 6 1026 64
NM_052933.3 NP_443165.1 4 - 5 631 64
GenBank mRNA AF294347.1 - 4 - 5 349 64
AF431815.1 - 4 - 5 631 64
AK093329.1 - 5 - 6 1025 64
BC038415.1 - 4 - 5 631 64

View Full Product Details