The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304968.1 | NP_001291897.1 | 6 - 7 | 1237 | 94 |
NM_052933.3 | NP_443165.1 | 5 - 6 | 842 | 94 | |
GenBank mRNA | AF294347.1 | - | 5 - 6 | 560 | 94 |
AF431815.1 | - | 5 - 6 | 842 | 94 | |
AK093329.1 | - | 6 - 7 | 1236 | 94 | |
BC038415.1 | - | 5 - 6 | 842 | 94 |