The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052947.3 | NP_443179.3 | 11 - 12 | 6460 | 86 |
XM_011525802.2 | XP_011524104.1 | 9 - 10 | 7080 | 86 | |
XM_017025535.1 | XP_016881024.1 | 11 - 12 | 6694 | 86 | |
XM_017025536.1 | XP_016881025.1 | 11 - 12 | 6694 | 86 | |
XM_017025538.1 | XP_016881027.1 | 8 - 9 | 5578 | 86 | |
XM_017025539.1 | XP_016881028.1 | 8 - 9 | 4326 | 86 | |
GenBank mRNA | AL832018.2 | - | 8 - 9 | 4792 | 86 |
AY044450.1 | - | 8 - 9 | 4527 | 86 | |
BX647446.1 | - | 7 - 8 | 3938 | 86 | |
BX647639.1 | - | 8 - 9 | 5326 | 86 |