The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052952.2 | NP_443184.1 | 1 - 2 | 244 | 84 |
GenBank mRNA | AY039011.1 | - | 1 - 2 | 245 | 84 |