Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VIT
Entrez Gene ID
5212
Gene Name
vitrin
Gene Aliases
-
Chromosome Location
Chr.2: 36696685 - 36814794 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177971.1 NP_001171442.1 8 - 9 1039 79
XM_011532906.1 XP_011531208.1 7 - 8 744 79
GenBank mRNA AF063833.1 - 8 - 9 863 79

View Full Product Details