The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057090.2 | NP_476431.2 | 2 - 3 | 714 | 85 |
NM_057091.2 | NP_476432.2 | 2 - 3 | 714 | 85 | |
GenBank mRNA | AF120274.1 | - | 2 - 3 | 714 | 85 |