Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL6A3
Entrez Gene ID
1293
Gene Name
collagen type VI alpha 3 chain
Gene Aliases
BTHLM1, DYT27, UCMD1
Chromosome Location
Chr.2: 237324012 - 237434822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004369.3 NP_004360.2 29 - 30 7217 72
NM_057166.4 NP_476507.3 26 - 27 5396 72
NM_057167.3 NP_476508.2 28 - 29 6599 72
XM_005246065.1 XP_005246122.1 28 - 29 6574 72
XM_005246066.1 XP_005246123.1 26 - 27 5720 72
XM_006712253.1 XP_006712316.1 29 - 30 6673 72
XM_011510574.1 XP_011508876.1 29 - 30 7171 72
XM_017003303.1 XP_016858792.1 34 - 35 7224 72
XM_017003304.1 XP_016858793.1 31 - 32 5436 72
GenBank mRNA AB209636.1 - 20 - 21 2508 72
BC033174.1 - 6 - 7 340 72
BC144595.1 - 26 - 27 5337 72
BC150625.1 - 26 - 27 5337 72
BC171790.1 - 26 - 27 5337 72
BX641155.1 - 6 - 7 1186 72
M20778.1 - 15 - 16 821 72
X52022.1 - 29 - 30 7187 72

View Full Product Details