The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_079834.2 | NP_524558.1 | 4 - 5 | 403 | 80 |
GenBank mRNA | AK074967.1 | - | 4 - 5 | 378 | 80 |
AK307641.1 | - | 3 - 4 | 329 | 80 | |
AK307700.1 | - | 3 - 4 | 327 | 80 | |
AL832731.1 | - | 2 - 3 | 232 | 80 | |
BC011747.2 | - | 4 - 5 | 365 | 80 | |
BC016509.1 | - | 4 - 5 | 377 | 80 | |
BC062598.1 | - | 4 - 5 | 376 | 80 |