The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080670.3 | NP_542401.1 | 2 - 3 | 388 | 80 |
GenBank mRNA | AF445025.1 | - | 1 - 2 | 1641 | 80 |
AK122601.1 | - | 2 - 3 | 125 | 80 | |
AK307282.1 | - | 2 - 3 | 388 | 80 | |
AL833972.1 | - | 1 - 2 | 107 | 80 | |
BC009906.2 | - | 2 - 3 | 118 | 80 |