Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GIMAP1
Entrez Gene ID
170575
Gene Name
GTPase, IMAP family member 1
Gene Aliases
HIMAP1, IAN2, IMAP1, IMAP38
Chromosome Location
Chr.7: 150716557 - 150724280 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130759.3 NP_570115.1 1 - 2 131 60
GenBank mRNA AJ306287.1 - 1 - 2 64 60
AK091818.1 - 1 - 2 78 60
AK315127.1 - 1 - 2 75 60
BC040736.1 - 1 - 2 59 60
DA817535.1 - 1 - 2 131 60

Gene Symbol
GIMAP1-GIMAP5
Entrez Gene ID
100527949
Gene Name
GIMAP1-GIMAP5 readthrough
Gene Aliases
-
Chromosome Location
Chr.7: 150716557 - 150743649 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199577.1 NP_001186506.1 1 - 2 131 60
NM_001303630.1 NP_001290559.1 1 - 2 131 60

View Full Product Details