The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278708.1 | NP_001265637.1 | 11 - 12 | 1450 | 72 |
NM_001278709.1 | NP_001265638.1 | 11 - 12 | 1439 | 72 | |
NM_002808.4 | NP_002799.3 | 13 - 14 | 1871 | 72 | |
GenBank mRNA | AB209843.1 | - | 13 - 14 | 1717 | 72 |
AK095245.1 | - | 13 - 14 | 2262 | 72 | |
AK297256.1 | - | 14 - 15 | 1702 | 72 | |
AK301759.1 | - | 11 - 12 | 1452 | 72 | |
AK302177.1 | - | 11 - 12 | 1439 | 72 | |
AK315474.1 | - | 13 - 14 | 1730 | 72 | |
AL162014.1 | - | 13 - 14 | 1734 | 72 | |
BC002368.2 | - | 13 - 14 | 1703 | 72 | |
BC002997.1 | - | 13 - 14 | 1724 | 72 | |
BC007897.2 | - | 13 - 14 | 1702 | 72 | |
BT009736.1 | - | 13 - 14 | 1702 | 72 | |
D78151.1 | - | 13 - 14 | 1735 | 72 | |
U12596.1 | - | 13 - 14 | 1640 | 72 | |
U18247.1 | - | 9 - 10 | 1099 | 72 | |
X86446.1 | - | 13 - 14 | 1675 | 72 |