Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MADD
Entrez Gene ID
8567
Gene Name
MAP kinase activating death domain
Gene Aliases
DENN, IG20, RAB3GEP
Chromosome Location
Chr.11: 47269376 - 47330031 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135943.1 NP_001129415.1 24 - 25 3952 82
NM_001135944.1 NP_001129416.1 24 - 25 3919 82
NM_003682.3 NP_003673.3 27 - 28 4237 82
NM_130470.2 NP_569826.2 24 - 25 4226 82
NM_130471.2 NP_569827.2 25 - 26 3991 82
NM_130472.2 NP_569828.2 24 - 25 3931 82
NM_130473.2 NP_569829.2 25 - 26 4120 82
NM_130474.2 NP_569830.2 24 - 25 3931 82
NM_130475.2 NP_569831.1 27 - 28 4261 82
NM_130476.2 NP_569832.2 24 - 25 4081 82
XM_005253189.2 XP_005253246.1 27 - 28 4261 82
XM_005253196.2 XP_005253253.1 25 - 26 4144 82
XM_005253199.2 XP_005253256.1 24 - 25 4084 82
XM_005253200.2 XP_005253257.1 27 - 28 4258 82
XM_005253201.2 XP_005253258.1 25 - 26 4015 82
XM_005253203.2 XP_005253260.1 24 - 25 3955 82
XM_005253204.2 XP_005253261.1 24 - 25 3943 82
XM_005253205.1 XP_005253262.1 24 - 25 3955 82
XM_017018478.1 XP_016873967.1 27 - 28 4249 82
XM_017018479.1 XP_016873968.1 26 - 27 4195 82
XM_017018480.1 XP_016873969.1 25 - 26 4141 82
XM_017018481.1 XP_016873970.1 25 - 26 4135 82
XM_017018482.1 XP_016873971.1 25 - 26 4132 82
XM_017018483.1 XP_016873972.1 27 - 28 4120 82
XM_017018484.1 XP_016873973.1 24 - 25 4072 82
XM_017018485.1 XP_016873974.1 24 - 25 4033 82
XM_017018486.1 XP_016873975.1 26 - 27 4066 82
XM_017018487.1 XP_016873976.1 26 - 27 4060 82
XM_017018488.1 XP_016873977.1 27 - 28 4249 82
XM_017018489.1 XP_016873978.1 27 - 28 4210 82
XM_017018490.1 XP_016873979.1 26 - 27 4194 82
XM_017018491.1 XP_016873980.1 26 - 27 4160 82
XM_017018492.1 XP_016873981.1 26 - 27 4195 82
XM_017018493.1 XP_016873982.1 26 - 27 4176 82
XM_017018494.1 XP_016873983.1 25 - 26 4147 82
XM_017018495.1 XP_016873984.1 25 - 26 4144 82
XM_017018496.1 XP_016873985.1 25 - 26 4091 82
XM_017018497.1 XP_016873986.1 25 - 26 4096 82
XM_017018498.1 XP_016873987.1 27 - 28 4120 82
XM_017018499.1 XP_016873988.1 27 - 28 4089 82
XM_017018500.1 XP_016873989.1 24 - 25 4081 82
XM_017018501.1 XP_016873990.1 26 - 27 4034 82
XM_017018502.1 XP_016873991.1 26 - 27 4047 82
XM_017018503.1 XP_016873992.1 26 - 27 4031 82
XM_017018504.1 XP_016873993.1 26 - 27 4044 82
XM_017018505.1 XP_016873994.1 26 - 27 4022 82
XM_017018506.1 XP_016873995.1 26 - 27 4027 82
XM_017018507.1 XP_016873996.1 26 - 27 4047 82
XM_017018508.1 XP_016873997.1 25 - 26 4018 82
XM_017018509.1 XP_016873998.1 25 - 26 3971 82
XM_017018510.1 XP_016873999.1 25 - 26 4006 82
GenBank mRNA AB002356.1 - 27 - 28 4233 82
AF440100.1 - 27 - 28 4211 82
AF440101.1 - 25 - 26 4094 82
AF440102.1 - 25 - 26 3965 82
AF440103.1 - 24 - 25 3905 82
AF440434.1 - 24 - 25 3905 82
AK292442.1 - 24 - 25 3883 82
AK308780.1 - 8 - 9 689 82
BC040484.1 - 24 - 25 3936 82
U44953.1 - 24 - 25 4045 82
U77352.1 - 24 - 25 4194 82

View Full Product Details