Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BDH1
Entrez Gene ID
622
Gene Name
3-hydroxybutyrate dehydrogenase, type 1
Gene Aliases
BDH, SDR9C1
Chromosome Location
Chr.3: 197509783 - 197573323 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004051.4 NP_004042.1 1 - 2 209 103
XM_017007010.1 XP_016862499.1 1 - 2 429 103
XM_017007012.1 XP_016862501.1 1 - 2 428 103
GenBank mRNA BC005844.2 - 1 - 2 173 103
BC011964.1 - 1 - 2 158 103
BC019317.1 - 1 - 2 173 103
BE782251.1 - - 270 103
BQ897235.1 - 1 - 2 206 103

View Full Product Details