The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004178.4 | NP_004169.3 | 4 - 5 | 443 | 136 |
NM_134323.1 | NP_599150.1 | 4 - 5 | 908 | 136 | |
NM_134324.2 | NP_599151.2 | 4 - 5 | 483 | 136 | |
XM_005269114.1 | XP_005269171.1 | 4 - 5 | 478 | 136 | |
XM_005269115.2 | XP_005269172.1 | 4 - 5 | 485 | 136 | |
XM_005269117.1 | XP_005269174.1 | 4 - 5 | 527 | 136 | |
XM_005269120.4 | XP_005269177.1 | 4 - 5 | 891 | 136 | |
XM_005269122.3 | XP_005269179.1 | 4 - 5 | 463 | 136 | |
XM_006719581.1 | XP_006719644.1 | 3 - 4 | 327 | 136 | |
XM_011538712.2 | XP_011537014.1 | 4 - 5 | 532 | 136 | |
XM_017019910.1 | XP_016875399.1 | 4 - 5 | 478 | 136 | |
XM_017019911.1 | XP_016875400.1 | 4 - 5 | 621 | 136 | |
GenBank mRNA | BC002419.2 | - | 4 - 5 | 891 | 136 |
BC005860.2 | - | 4 - 5 | 773 | 136 | |
BI771121.1 | - | 4 - 5 | 486 | 136 | |
BT007140.1 | - | 4 - 5 | 420 | 136 | |
M60801.1 | - | 4 - 5 | 136 | ||
U08998.2 | - | 4 - 5 | 508 | 136 |