The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004178.4 | NP_004169.3 | 6 - 7 | 631 | 90 |
NM_134323.1 | NP_599150.1 | 6 - 7 | 1096 | 90 | |
NM_134324.2 | NP_599151.2 | 6 - 7 | 671 | 90 | |
XM_005269114.1 | XP_005269171.1 | 6 - 7 | 666 | 90 | |
XM_005269115.2 | XP_005269172.1 | 6 - 7 | 673 | 90 | |
XM_005269120.4 | XP_005269177.1 | 6 - 7 | 1079 | 90 | |
XM_005269122.3 | XP_005269179.1 | 6 - 7 | 651 | 90 | |
XM_006719581.1 | XP_006719644.1 | 5 - 6 | 515 | 90 | |
XM_011538712.2 | XP_011537014.1 | 6 - 7 | 720 | 90 | |
XM_017019911.1 | XP_016875400.1 | 6 - 7 | 809 | 90 | |
GenBank mRNA | AK290737.1 | - | 6 - 7 | 669 | 90 |
BC002419.2 | - | 6 - 7 | 1079 | 90 | |
BC005860.2 | - | 6 - 7 | 961 | 90 | |
BT007140.1 | - | 6 - 7 | 608 | 90 | |
M60801.1 | - | 6 - 7 | 599 | 90 | |
U08998.2 | - | 6 - 7 | 696 | 90 |