The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145146.1 | NP_001138618.1 | 9 - 10 | 1123 | 61 |
NM_001145147.1 | NP_001138619.1 | 7 - 8 | 916 | 61 | |
NM_001145148.1 | NP_001138620.1 | 8 - 9 | 1036 | 61 | |
NM_001303018.1 | NP_001289947.1 | 6 - 7 | 830 | 61 | |
NM_001303020.1 | NP_001289949.1 | 7 - 8 | 950 | 61 | |
NM_004382.4 | NP_004373.2 | 8 - 9 | 1036 | 61 | |
GenBank mRNA | AB451466.1 | - | 8 - 9 | 771 | 61 |
AF180301.1 | - | 8 - 9 | 771 | 61 | |
AY457172.1 | - | 8 - 9 | 771 | 61 | |
BC096836.1 | - | 8 - 9 | 791 | 61 | |
EU012435.1 | - | 7 - 8 | 685 | 61 | |
EU176016.1 | - | 5 - 6 | 468 | 61 | |
FJ200483.1 | - | 6 - 7 | 565 | 61 | |
HM852949.1 | - | 9 - 10 | 1093 | 61 | |
L23332.1 | - | 8 - 9 | 808 | 61 | |
L23333.1 | - | 9 - 10 | 858 | 61 | |
U16273.1 | - | 7 - 8 | 669 | 61 | |
X72304.1 | - | 8 - 9 | 997 | 61 |