The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001314042.1 | NP_001300971.1 | 14 - 15 | 3097 | 82 |
NM_006618.4 | NP_006609.3 | 13 - 14 | 2989 | 82 | |
XM_011509088.2 | XP_011507390.1 | 12 - 13 | 1906 | 82 | |
XM_011509090.2 | XP_011507392.1 | 14 - 15 | 2078 | 82 | |
XM_011509091.2 | XP_011507393.1 | 14 - 15 | 1815 | 82 | |
XM_011509092.2 | XP_011507394.1 | 13 - 14 | 1932 | 82 | |
GenBank mRNA | AF087481.1 | - | 14 - 15 | 2048 | 82 |
AJ132440.1 | - | 13 - 14 | 1913 | 82 | |
AJ243706.1 | - | 13 - 14 | 2992 | 82 | |
AK022553.1 | - | 4 - 5 | 639 | 82 | |
AK122752.1 | - | 13 - 14 | 1943 | 82 | |
AL133040.1 | - | 9 - 10 | 1242 | 82 | |
AL133048.1 | - | 2 - 3 | 278 | 82 |