Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NT5C2
Entrez Gene ID
22978
Gene Name
5'-nucleotidase, cytosolic II
Gene Aliases
GMP, NT5B, PNT5, SPG45, SPG65, cN-II
Chromosome Location
Chr.10: 103088017 - 103193306 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134373.2 NP_001127845.1 11 - 12 911 67
NM_012229.4 NP_036361.1 12 - 13 1009 67
XM_005269632.4 XP_005269689.1 12 - 13 920 67
XM_005269633.4 XP_005269690.1 13 - 14 1013 67
XM_005269634.4 XP_005269691.1 13 - 14 1045 67
XM_005269635.4 XP_005269692.1 12 - 13 1235 67
XM_005269636.4 XP_005269693.1 12 - 13 1021 67
XM_005269637.4 XP_005269694.1 11 - 12 1739 67
XM_005269638.4 XP_005269695.1 11 - 12 3040 67
XM_005269639.4 XP_005269696.1 10 - 11 764 67
XM_005269640.4 XP_005269697.1 13 - 14 1036 67
XM_005269641.4 XP_005269698.1 12 - 13 938 67
XM_005269642.4 XP_005269699.1 11 - 12 863 67
XM_005269643.4 XP_005269700.1 10 - 11 803 67
XM_005269644.4 XP_005269701.1 14 - 15 1060 67
XM_005269645.4 XP_005269702.1 13 - 14 962 67
XM_005269646.4 XP_005269703.1 12 - 13 888 67
XM_006717721.3 XP_006717784.1 12 - 13 962 67
XM_006717723.3 XP_006717786.1 10 - 11 759 67
XM_011539537.2 XP_011537839.1 13 - 14 1045 67
XM_017015947.1 XP_016871436.1 10 - 11 789 67
XM_017015948.1 XP_016871437.1 13 - 14 1145 67
XM_017015949.1 XP_016871438.1 13 - 14 1130 67
XM_017015950.1 XP_016871439.1 11 - 12 846 67
XM_017015951.1 XP_016871440.1 10 - 11 772 67
XM_017015952.1 XP_016871441.1 12 - 13 1069 67
XM_017015953.1 XP_016871442.1 13 - 14 1464 67
XM_017015954.1 XP_016871443.1 12 - 13 1403 67
XM_017015955.1 XP_016871444.1 12 - 13 1090 67
XM_017015956.1 XP_016871445.1 12 - 13 1296 67
XM_017015957.1 XP_016871446.1 14 - 15 1154 67
XM_017015958.1 XP_016871447.1 15 - 16 1273 67
XM_017015959.1 XP_016871448.1 11 - 12 864 67
XM_017015960.1 XP_016871449.1 12 - 13 827 67
XM_017015961.1 XP_016871450.1 12 - 13 1222 67
XM_017015962.1 XP_016871451.1 12 - 13 1056 67
XM_017015963.1 XP_016871452.1 12 - 13 1160 67
XM_017015964.1 XP_016871453.1 12 - 13 1139 67
XM_017015965.1 XP_016871454.1 11 - 12 1016 67
XM_017015966.1 XP_016871455.1 11 - 12 1016 67
XM_017015967.1 XP_016871456.1 11 - 12 951 67
XM_017015968.1 XP_016871457.1 12 - 13 944 67
XM_017015969.1 XP_016871458.1 12 - 13 1038 67
XM_017015970.1 XP_016871459.1 10 - 11 1681 67
XM_017015971.1 XP_016871460.1 9 - 10 725 67
XM_017015972.1 XP_016871461.1 13 - 14 979 67
XM_017015973.1 XP_016871462.1 14 - 15 1003 67
XM_017015974.1 XP_016871463.1 13 - 14 1003 67
XM_017015975.1 XP_016871464.1 11 - 12 844 67
XM_017015976.1 XP_016871465.1 11 - 12 850 67
GenBank mRNA AK127670.1 - 12 - 13 1000 67
AK291667.1 - 11 - 12 877 67
AK295593.1 - 10 - 11 1053 67
AK312517.1 - 11 - 12 877 67
BC001595.1 - 11 - 12 960 67
BX648078.1 - 13 - 14 2811 67
D38524.1 - 11 - 12 899 67

View Full Product Details