Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL37
Entrez Gene ID
27178
Gene Name
interleukin 37
Gene Aliases
FIL1, FIL1(ZETA), FIL1Z, IL-1F7, IL-1H, IL-1H4, IL-1RP1, IL-37, IL1F7, IL1H4, IL1RP1
Chromosome Location
Chr.2: 112908889 - 112918882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014439.3 NP_055254.2 4 - 5 453 60
NM_173202.1 NP_775294.1 3 - 4 390 60
NM_173203.1 NP_775295.1 2 - 3 270 60
NM_173204.1 NP_775296.1 3 - 4 333 60
NM_173205.1 NP_775297.1 3 - 4 333 60
XM_011510963.2 XP_011509265.1 5 - 6 2454 60
XM_011510964.2 XP_011509266.1 4 - 5 2701 60
GenBank mRNA AB451333.1 - 3 - 4 348 60
AB451478.1 - 3 - 4 348 60
AF167368.1 - 4 - 5 453 60
AF200496.1 - 4 - 5 459 60
AF201832.1 - 3 - 4 333 60
AF251118.1 - 4 - 5 483 60
AF251119.1 - 4 - 5 411 60
AF251120.1 - 3 - 4 291 60
AY071840.1 - 3 - 4 348 60
AY071841.1 - 2 - 3 228 60
BC020637.1 - 4 - 5 414 60

View Full Product Details