The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128852.1 | NP_001122324.1 | 13 - 14 | 1911 | 106 |
NM_001128853.1 | NP_001122325.1 | 13 - 14 | 1914 | 106 | |
NM_001128854.1 | NP_001122326.1 | 13 - 14 | 1911 | 106 | |
NM_015908.5 | NP_056992.4 | 13 - 14 | 1914 | 106 | |
XM_005250405.2 | XP_005250462.1 | 13 - 14 | 1771 | 106 | |
XM_005250406.2 | XP_005250463.1 | 13 - 14 | 1768 | 106 | |
XM_005250407.2 | XP_005250464.1 | 13 - 14 | 1771 | 106 | |
XM_005250408.2 | XP_005250465.1 | 13 - 14 | 1768 | 106 | |
XM_017012290.1 | XP_016867779.1 | 14 - 15 | 2096 | 106 | |
XM_017012291.1 | XP_016867780.1 | 14 - 15 | 2093 | 106 | |
GenBank mRNA | AF082871.1 | - | 3 - 4 | 322 | 106 |
AF248955.1 | - | 13 - 14 | 1877 | 106 | |
AK296131.1 | - | 7 - 8 | 880 | 106 | |
AK302820.1 | - | 13 - 14 | 1803 | 106 | |
AK311728.1 | - | 5 - 6 | 742 | 106 | |
AL096723.1 | - | 13 - 14 | 1865 | 106 | |
BC000082.2 | - | 4 - 5 | 435 | 106 | |
BC069249.1 | - | 11 - 12 | 1455 | 106 | |
BC109117.1 | - | 13 - 14 | 1805 | 106 | |
BQ059529.1 | - | 1 - 2 | 210 | 106 |