The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015986.3 | NP_057070.3 | 2 - 3 | 448 | 61 |
XM_011524879.1 | XP_011523181.1 | 2 - 3 | 622 | 61 | |
GenBank mRNA | AF046059.1 | - | 2 - 3 | 361 | 61 |
AF120151.1 | - | - | 66 | 61 | |
AK313315.1 | - | 2 - 3 | 437 | 61 | |
BC023567.2 | - | 2 - 3 | 370 | 61 | |
BU902521.1 | - | 2 - 3 | 489 | 61 | |
DQ298450.1 | - | 2 - 3 | 327 | 61 |