Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAV2
Entrez Gene ID
89797
Gene Name
neuron navigator 2
Gene Aliases
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2
Chromosome Location
Chr.11: 19345200 - 20121601 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111018.1 NP_001104488.1 37 - 38 7206 56
NM_001111019.2 NP_001104489.1 27 - 28 4618 56
NM_001244963.1 NP_001231892.1 40 - 41 7707 56
NM_145117.4 NP_660093.2 37 - 38 7530 56
NM_182964.5 NP_892009.3 38 - 39 7539 56
XM_005253214.4 XP_005253271.1 26 - 27 9014 56
XM_006718364.3 XP_006718427.1 39 - 40 7960 56
XM_006718365.3 XP_006718428.1 39 - 40 7930 56
XM_006718366.3 XP_006718429.1 40 - 41 7193 56
XM_006718367.3 XP_006718430.1 29 - 30 9227 56
XM_006718368.3 XP_006718431.1 26 - 27 9050 56
XM_006718369.2 XP_006718432.1 29 - 30 4722 56
XM_011520444.2 XP_011518746.1 39 - 40 8020 56
XM_011520445.2 XP_011518747.1 39 - 40 7963 56
XM_011520446.2 XP_011518748.1 38 - 39 7921 56
XM_011520447.2 XP_011518749.1 38 - 39 7918 56
XM_011520448.2 XP_011518750.1 40 - 41 7900 56
XM_011520449.2 XP_011518751.1 37 - 38 7855 56
XM_011520450.2 XP_011518752.1 37 - 38 7819 56
XM_011520451.2 XP_011518753.1 39 - 40 7407 56
XM_011520452.2 XP_011518754.1 38 - 39 7669 56
XM_011520453.2 XP_011518755.1 29 - 30 4657 56
XM_011520454.2 XP_011518756.1 26 - 27 4447 56
XM_017018520.1 XP_016874009.1 41 - 42 7821 56
XM_017018521.1 XP_016874010.1 36 - 37 7810 56
XM_017018522.1 XP_016874011.1 40 - 41 7777 56
XM_017018523.1 XP_016874012.1 40 - 41 8706 56
XM_017018524.1 XP_016874013.1 40 - 41 7529 56
XM_017018525.1 XP_016874014.1 34 - 35 6372 56
XM_017018526.1 XP_016874015.1 30 - 31 7008 56
GenBank mRNA AB037840.3 - 38 - 39 7540 56
AB063115.1 - 37 - 38 7206 56
AB063116.1 - 37 - 38 7499 56
AF466143.1 - 37 - 38 7245 56
AF466144.1 - 38 - 39 7254 56
AJ488102.1 - 40 - 41 7445 56
AK001892.1 - 12 - 13 1742 56
AK074287.1 - 27 - 28 4617 56
AK096037.1 - 3 - 4 1845 56
AK290458.1 - 21 - 22 2820 56
BC016054.1 - 19 - 20 2523 56
BC150290.1 - 38 - 39 7540 56

View Full Product Details