The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017922.1 | NP_001017922.1 | 3 - 4 | 353 | 67 |
NM_018538.3 | NP_061008.2 | 2 - 3 | 249 | 67 | |
XM_006710313.3 | XP_006710376.1 | 4 - 5 | 618 | 67 | |
XM_011540570.2 | XP_011538872.1 | 3 - 4 | 309 | 67 | |
XM_011540571.2 | XP_011538873.1 | 3 - 4 | 337 | 67 | |
GenBank mRNA | AF311285.1 | - | 2 - 3 | 249 | 67 |
AK056138.1 | - | 3 - 4 | 67 | ||
AK295704.1 | - | 3 - 4 | 271 | 67 | |
AK314436.1 | - | 3 - 4 | 337 | 67 | |
AY049028.1 | - | 2 - 3 | 267 | 67 | |
BC099703.1 | - | 2 - 3 | 182 | 67 | |
BC099707.1 | - | 2 - 3 | 187 | 67 | |
BC099712.1 | - | 2 - 3 | 182 | 67 | |
BC099713.1 | - | 2 - 3 | 182 | 67 | |
BX507568.1 | - | 3 - 4 | 353 | 67 |