The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019034.2 | NP_061907.2 | 2 - 3 | 281 | 61 |
GenBank mRNA | AF239923.1 | - | 2 - 3 | 255 | 61 |
AK000254.1 | - | 2 - 3 | 247 | 61 | |
AK289482.1 | - | 2 - 3 | 855 | 61 | |
BC018208.1 | - | 2 - 3 | 260 | 61 | |
BQ053707.1 | - | 2 - 3 | 262 | 61 | |
BU527972.1 | - | 2 - 3 | 263 | 61 |