The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017396.2 | NP_001017396.1 | 2 - 3 | 564 | 93 |
NM_001291605.1 | NP_001278534.1 | 3 - 4 | 636 | 93 | |
NM_021088.3 | NP_066574.2 | 3 - 4 | 636 | 93 | |
GenBank mRNA | AK055072.1 | - | 3 - 4 | 615 | 93 |
AK290798.1 | - | 2 - 3 | 174 | 93 | |
AK299058.1 | - | 3 - 4 | 236 | 93 | |
AK315762.1 | - | 3 - 4 | 248 | 93 | |
AK315829.1 | - | 2 - 3 | 169 | 93 | |
BC005068.1 | - | 3 - 4 | 608 | 93 | |
X60152.1 | - | 2 - 3 | 549 | 93 | |
X60154.1 | - | 3 - 4 | 225 | 93 |