The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193314.1 | NP_001180243.1 | 8 - 9 | 1149 | 68 |
NM_001193315.1 | NP_001180244.1 | 8 - 9 | 749 | 68 | |
NM_001193316.1 | NP_001180245.1 | 7 - 8 | 602 | 68 | |
NM_001193317.1 | NP_001180246.1 | 8 - 9 | 926 | 68 | |
NM_022067.3 | NP_071350.2 | 9 - 10 | 1020 | 68 | |
XM_017021580.1 | XP_016877069.1 | 8 - 9 | 740 | 68 | |
XM_017021581.1 | XP_016877070.1 | 8 - 9 | 874 | 68 | |
GenBank mRNA | AK022769.1 | - | 8 - 9 | 697 | 68 |
AK022925.1 | - | 8 - 9 | 675 | 68 | |
AK098254.1 | - | 8 - 9 | 1110 | 68 | |
AK222706.1 | - | 8 - 9 | 695 | 68 | |
AK297388.1 | - | 8 - 9 | 851 | 68 | |
AK298354.1 | - | 7 - 8 | 548 | 68 | |
BC015054.1 | - | 9 - 10 | 941 | 68 | |
CR457294.1 | - | 7 - 8 | 603 | 68 |