Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRNP25
Entrez Gene ID
79622
Gene Name
small nuclear ribonucleoprotein U11/U12 subunit 25
Gene Aliases
C16orf33
Chromosome Location
Chr.16: 53829 - 57669 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024571.3 NP_078847.1 2 - 3 320 59
GenBank mRNA AK026593.1 - 2 - 3 289 59
BC001381.1 - 2 - 3 1142 59
BC009179.1 - 2 - 3 271 59
Z84714.1 - 2 - 3 234 59

View Full Product Details