Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHO2
Entrez Gene ID
80301
Gene Name
pleckstrin homology domain containing O2
Gene Aliases
PLEKHQ1, PP1628, pp9099
Chromosome Location
Chr.15: 64841883 - 64868002 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195059.1 NP_001181988.1 2 - 3 269 84
NM_025201.4 NP_079477.2 3 - 4 419 84
GenBank mRNA AF494534.1 - 3 - 4 368 84
AK096203.1 - 3 - 4 387 84
BC008744.2 - 2 - 3 230 84

View Full Product Details