The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159286.1 | NP_001152758.1 | 16 - 17 | 1966 | 74 |
NM_032019.5 | NP_114408.3 | 17 - 18 | 2026 | 74 | |
GenBank mRNA | AF393962.1 | - | 17 - 18 | 1653 | 74 |
AF407272.1 | - | 17 - 18 | 1653 | 74 | |
AF407273.1 | - | 16 - 17 | 1593 | 74 | |
AF426160.1 | - | 17 - 18 | 2006 | 74 | |
AY450395.1 | - | 14 - 15 | 1418 | 74 | |
BC032049.1 | - | 17 - 18 | 1839 | 74 | |
BC094734.1 | - | 15 - 16 | 1694 | 74 | |
BC125082.1 | - | 11 - 12 | 1175 | 74 | |
BC125083.1 | - | 17 - 18 | 1700 | 74 | |
CR456465.1 | - | 17 - 18 | 1709 | 74 |